missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFAND2B Monoclonal Antibody (OTI3A5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502441S
This item is not returnable.
View return policy
Description
ZFAND2B Monoclonal antibody specifically detects ZFAND2B in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| ZFAND2B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WV99 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 130617 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Western Blot | |
| OTI3A5 | |
| Unconjugated | |
| ZFAND2B | |
| 1110060O18Rik; AIRAPL; AIRAP-like protein; AN1-type zinc finger protein 2B; arsenite-inducible RNA-associated protein-like protein; C81256; ZFAND2B; zinc finger AN1-type containing 2B; zinc finger, AN1 type domain 2B; zinc finger, AN1-type domain 2B | |
| Full length human recombinant protein of human ZFAND2B produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction