missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFAND5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93657-0.02ml
This item is not returnable.
View return policy
Description
ZFAND5 Polyclonal antibody specifically detects ZFAND5 in Mouse samples. It is validated for Western Blot
Specifications
| ZFAND5 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| Zinc finger A20 domain-containing protein 2, Zinc finger protein 216ZA20D2, zinc finger, AN1-type domain 5, ZNF216A20 domain containing 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-150 of human ZFAND5 (NP_001095890.1). RQQNSGRMSPMGTASGSNSPTSDSASVQRADTSLNNCEGAAGSTSEKSRNVPVAALPVTQQMTEMSISREDKITTPKTEVSEPVVTQPSPSVSQPSTSQSEEKAPELPKPK | |
| 0.02 mL | |
| Apoptosis, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 7763 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction