missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFAND5 Antibody (OTI5H8), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74922B
This item is not returnable.
View return policy
Description
ZFAND5 Monoclonal specifically detects ZFAND5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ZFAND5 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| ZFAND5 | |
| Full length recombinant protein of human ZFAND5 (NP_005998) produced in E.coli. | |
| 0.1 mL | |
| 7763.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI5H8 | |
| Western Blot, Immunohistochemistry | |
| Zinc finger A20 domain-containing protein 2, Zinc finger protein 216ZA20D2, zinc finger, AN1-type domain 5, ZNF216A20 domain containing 2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction