missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFAND5 Monoclonal Antibody (OTI1D8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800482S
This item is not returnable.
View return policy
Description
ZFAND5 Monoclonal antibody specifically detects ZFAND5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| ZFAND5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O76080 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7763 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D8 | |
| Unconjugated | |
| ZFAND5 | |
| 2310057A04Rik; 5830475F03Rik; AA415485; AN1-type zinc finger protein 5; EST00848 homolog; ubiquitin homolog; Za20d2; Zfand5; ZFAND5A; Zfp216; Zinc finger A20 domain-containing protein 2; zinc finger AN1-type containing 5; zinc finger protein 216; zinc finger, A20 domain containing 2; zinc finger, AN1-type domain 5; ZNF216 | |
| Full length human recombinant protein of human ZFAND5 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction