missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFP14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569707
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Dog: 79%; Horse: 86%; Human: 100%; Rat: 79%.
ZFP14 may be involved in transcriptional regulation.
Specifications
| ZFP14 | |
| Polyclonal | |
| Unconjugated | |
| ZFP14 | |
| KIAA1559; mouse zinc finger protein 14-like; ZFP14; Zfp-14; ZFP14 zinc finger protein; Zfp968; zinc finger protein 14 homolog; zinc finger protein 2; zinc finger protein 531; zinc finger protein 968; ZNF531 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57677 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9HCL3 | |
| ZFP14 | |
| synthetic peptide directed towards the N terminal region of human ZFP14. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction