missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551175
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total ZFP2 protein.
Probable transcription factor involved in neuronal differentiation and/or phenotypic maintenance.
Specifications
| ZFP2 | |
| Polyclonal | |
| Unconjugated | |
| ZFP2 | |
| 9930007F06Rik; Fnp-2; Mkr2; mkr-2; mszf87; Protein mKR2; Zfp2; zfp-2; ZFP2 zinc finger protein; zinc finger protein 2; Zinc finger protein 2 homolog; zinc finger protein 751; ZNF751 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80108 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q6ZN57 | |
| ZFP2 | |
| Fusion protein of human ZFP2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction