missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zfp219 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76550UV
This item is not returnable.
View return policy
Description
Zfp219 Polyclonal antibody specifically detects Zfp219 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Zfp219 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 51222 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| FLJ14457, MGC126229, MGC126230, zinc finger protein 514 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human ZFP219. The immunogen is located within amino acids 90 - 140 of ZFP219. Amino Acid Squence: SHLRTHQPERPRSPAARL | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction