missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566074
This item is not returnable.
View return policy
Description
Immunogen sequence: GLMIFKKSPSS EKDRENNESE RGCSPSPNLV THQGDTTEGV SAFATSGQNF LEILESNKTQ RSSVGE Highest antigen sequence identity to the following orthologs - mouse 48%, rat 53%.
May be involved in transcriptional regulation.
Specifications
| ZFP3 | |
| Polyclonal | |
| Unconjugated | |
| ZFP3 | |
| ZFP3; Zfp-3; ZFP3 zinc finger protein; zinc finger protein 3 homolog; Zinc finger protein 752; zinc finger protein homologous to Zfp-3 in mouse; zinc finger protein-3; ZNF752 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 124961 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96NJ6 | |
| ZFP3 | |
| Recombinant Human ZFP3. Recombinant protein control fragment (Product #RP-106765). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction