missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZFP36 Monoclonal Antibody (K94030_9A2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556084
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: DLAAPGHPPV LRQSISFSGL PSGRRTSPPP PGLAGPSLSS SSFSPSSSPP PPGDLPLSPS AFSAAPGTPL ARRDPTPVCC PSCRRATPIS VWGPLGGLVR TPSVQSLGSD PDEYASSGSS LGGSDSPVFE AGVFAPPQPV
ZFP36 is a probable regulatory protein with a novel zinc finger structure involved in regulating the response to growth factors. Knockdown of ZFP36 increased both cognate macrophage gene mRNAs and inflammatory tumor necrosis factor protein release. Overexpression of ZFP36 resulted in decreased levels of the same genes supporting its role in regulating macrophage gene expression. Multiple phosphorylation sites in ZFP36 in mammalian cells were reported. ZFP36 can be phosphorylated by JNK as well as by the other members of the greater MAP kinase family. Gene expression profiling predicts clinical outcome of prostate cancer. Inappropriate ZFP36 production may be one factor that contributes to higher rheumatoid arthritis disease activity.
Specifications
| ZFP36 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P26651 | |
| Zfp36 | |
| Recombinant human ZFP36 (immunogen range is 171-310). | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunoprecipitation, Western Blot | |
| K94030_9A2 | |
| Unconjugated | |
| Zfp36 | |
| G0/G1 switch regulatory protein 24; G0S24; GOS24; growth factor-inducible nuclear protein NUP475; mRNA decay activator protein ZFP36; Nup475; RNF162A; Tax interacting protein; TIS11; TIS11A; TIS11D; TISII; TNF alpha inhibitor; TPA-induced sequence 11; Tristetraprolin; tristetraprolin; LOW QUALITY PROTEIN: mRNA decay activator protein ZFP36; tristetraproline; TTP; Zfp36; zfp-36; ZFP36 ring finger protein; zinc finger protein 36; zinc finger protein 36 homolog; zinc finger protein 36, C3H type, homolog; zinc finger protein, C3H type, 36 homolog | |
| Mouse | |
| Protein A | |
| RUO | |
| 7538 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction