missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP36 Monoclonal Antibody (OTI3C3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502379S
This item is not returnable.
View return policy
Description
ZFP36 Monoclonal antibody specifically detects ZFP36 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| ZFP36 | |
| Monoclonal | |
| 0.36 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P26651 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7538 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI3C3 | |
| Unconjugated | |
| Zfp36 | |
| G0/G1 switch regulatory protein 24; G0S24; GOS24; growth factor-inducible nuclear protein NUP475; mRNA decay activator protein ZFP36; Nup475; RNF162A; Tax interacting protein; TIS11; TIS11A; TIS11D; TISII; TNF alpha inhibitor; TPA-induced sequence 11; Tristetraprolin; tristetraprolin; LOW QUALITY PROTEIN: mRNA decay activator protein ZFP36; tristetraproline; TTP; Zfp36; zfp-36; ZFP36 ring finger protein; zinc finger protein 36; zinc finger protein 36 homolog; zinc finger protein 36, C3H type, homolog; zinc finger protein, C3H type, 36 homolog | |
| Full length human recombinant protein of human ZFP36 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction