missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP36 Monoclonal Antibody (OTI4E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502385
This item is not returnable.
View return policy
Description
ZFP36 is a probable regulatory protein with a novel zinc finger structure involved in regulating the response to growth factors. Knockdown of ZFP36 increased both cognate macrophage gene mRNAs and inflammatory tumor necrosis factor protein release. Overexpression of ZFP36 resulted in decreased levels of the same genes supporting its role in regulating macrophage gene expression. Multiple phosphorylation sites in ZFP36 in mammalian cells were reported. ZFP36 can be phosphorylated by JNK as well as by the other members of the greater MAP kinase family. Gene expression profiling predicts clinical outcome of prostate cancer. Inappropriate ZFP36 production may be one factor that contributes to higher rheumatoid arthritis disease activity.Specifications
| ZFP36 | |
| Monoclonal | |
| 0.55 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P26651 | |
| ZFP36 | |
| Full length human recombit protein of human ZFP36 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI4E1 | |
| Unconjugated | |
| ZFP36 | |
| G0S24, GOS24, NUP475, RNF162A, TIS11, TTP, zfp-36 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7538 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction