missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP36L1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94828-0.1ml
This item is not returnable.
View return policy
Description
ZFP36L1 Polyclonal antibody specifically detects ZFP36L1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ZFP36L1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| ZBRK1KRAB zinc finger protein ZFQR, ZFQR, Zinc finger and BRCA1-interacting protein with a KRAB domain 1, zinc finger protein 350, Zinc finger protein ZBRK1, zinc-finger protein ZBRK1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-338 of human ZFP36L1 (NP_004917.2). MTTTLVSATIFDLSEVLCKGNKMLNYSAPSAGGCLLDRKAVGTPAGGGFPRRHSVTLPSSKFHQNQLLSSLKGEPAPALSSRDSRFRDRSFSEGGERLLPTQKQPGGGQVNSSRYKTELCRPFEENGACKYGDKCQFAHGIHELRSLTRHPKYKTELCRTFHTIGFCPYGPRCHFIHNAEERRALAGARDLSADRPRLQHSFSFAGFPSAAATAAATGLLDSPTSITPPPILSADDLLGSPTLPDGTNNPFAFSSQELASLFAPSMGLPGGGSPTTFLFRPMSESPHMFDSPPSPQDSLSDQEGYLSSSSSSHSGSDSPTLDNSRRLPIFSRLSISDD | |
| 0.1 mL | |
| Cell Biology, Signal Transduction | |
| 677 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction