missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP37 Monoclonal Antibody (OTI5E3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810210
This item is not returnable.
View return policy
Description
ZFP37 may be involved in transcriptional regulation.Specifications
| ZFP37 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y6Q3 | |
| ZFP37 | |
| Full length human recombit protein of human ZFP37 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5E3 | |
| Unconjugated | |
| ZFP37 | |
| ZNF906, zfp-37 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7539 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?