Learn More
ZFP37 Monoclonal Antibody (OTI5E3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA810210S
Encompass_Preferred
 View more versions of this product
Product Code. 17593124
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
ZFP37 Monoclonal antibody specifically detects ZFP37 in Human samples. It is validated for Western Blot
Specifications
Specifications
ZFP37
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9Y6Q3
Mouse
Affinity chromatography
RUO
7539
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI5E3
Unconjugated
ZFP37
LOW QUALITY PROTEIN: zinc finger protein 37 homolog; male germ cell-specific zinc finger protein; Tzn; ZFP37; Zfp-37; ZFP37 zinc finger protein; zinc finger protein 37; Zinc finger protein 37 homolog; zinc finger protein homologous to Zfp37 in mouse; ZNF906
Full length human recombinant protein of human ZFP37 produced in E.coli.
30 μL