missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP42 Monoclonal Antibody (OTI3H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807532
This item is not returnable.
View return policy
Description
Rex1 is an exonuclease domain-containing transcription factor protein that can bind to Elongin. The Elongin complex stimulates the rate of transcription elongation by RNA polymerase II through suppression of transient pauses of the polymerase at various sites along the DNA template. Rex1 is a putative stem cell marker that is highly expressed in embryonic and adult stem cells.Specifications
| ZFP42 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96MM3 | |
| ZFP42 | |
| Human recombit protein fragment corresponding to amino acids 1-235 of human ZFP42 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3H9 | |
| Unconjugated | |
| ZFP42 | |
| REX-1, REX1, ZNF754, zfp-42 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 132625 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction