Learn More
Invitrogen™ Zfp472 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA569041
Product Code. 15847453
  • € 520.00 / 100µL
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Horse: 100%; Human: 100%; Mouse: 92%; Pig: 100%; Rat: 93%.
Zfp472 gene ontology annotations related to this gene include DNA-binding transcription factor activity; metal ion binding; nucleic acid binding.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Zfp472
Polyclonal
Unconjugated
Zfp472
KRAB box containing zinc finger protein; Krab box proteins interacting with myc 1; Krim1; KRIM-1; Krim-1A; Krim-1B; Zfp472; zin finger protein 472; zinc finger protein 472
Rabbit
Affinity Chromatography
RUO
224691
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
0
Zfp472
synthetic peptide directed towards the middle region of human Zfp472.