missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP57 Antibody (865), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-43813
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
ZFP57 Monoclonal specifically detects ZFP57 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ZFP57 | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| ZFP57 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human ZFP57. The exact sequence is proprietary. | |
| Protein A purified | |
| RUO | |
| 346171 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 865 | |
| Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| C6orf40, TNDM1, zfp-57, zinc finger protein 57 homolog (mouse), zinc finger protein 698, ZNF698 | |
| Mouse | |
| 52 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction