missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP64 Antibody (PCRP-ZFP64-1H2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08648F
This item is not returnable.
View return policy
Description
ZFP64 Monoclonal antibody specifically detects ZFP64 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
Specifications
| ZFP64 | |
| Monoclonal | |
| FITC | |
| dJ548G19.1, dJ831D17.1, FLJ10734, FLJ12628, Zfp-64, Zinc finger protein 338MGC940, zinc finger protein 64, zinc finger protein 64 homolog (mouse), ZNF338 | |
| Recombinant full-length human ZFP64 protein (Uniprot: Q9NTW7) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PCRP-ZFP64-1H2 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 55734 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction