Learn More
Invitrogen™ ZFP64 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590771
Description
Immunogen sequence: MNASSEGESF AGSVQIPGGT TVLVELTPDI HICGICKQQF NNLDAFVAHK QSGCQLTGTS AAAPSTVQFV SEETVPATQT QTTTRTITSE TQTITVSAPE FVFEHGYQTY LPTESNENQT ATVISLPAKS RTKKPTTPPA QKRLNCCYPG CQFKTAYGMK DMERHLKIHT; Positive Samples: 293T, NIH/3T3, Raji; Cellular Location: Nucleus.
ZFP64 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 9 C2H2-type zinc fingers. ZFP64 may be involved in transcriptional regulation.
Specifications
| ZFP64 | |
| Polyclonal | |
| Unconjugated | |
| ZFP64 | |
| ZFP64; Zfp-64; ZFP64 zinc finger protein; zinc finger protein 338; Zinc finger protein 64; zinc finger protein 64 homolog; ZNF338 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22722, 55734 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q99KE8, Q9NTW7 | |
| ZFP64 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human ZFP64 (NP_0606672). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.