missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP90 Antibody (PCRP-ZFP90-1C5), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24072AF488
This item is not returnable.
View return policy
Description
ZFP90 Monoclonal antibody specifically detects ZFP90 in Human samples. It is validated for Flow Cytometry
Specifications
| ZFP90 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| KIAA1954, NK10, zfp-90, zinc finger protein 476, zinc finger protein 756, zinc finger protein 90 homolog, zinc finger protein 90 homolog (mouse), ZNF756 | |
| Recombinant fragment corresponding to protein domain of ZFP90 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| PCRP-ZFP90-1C5 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 146198 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction