missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP90 Antibody (PCRP-ZFP90-1C5), Novus Biologicals™
Mouse Monoclonal Antibody
€ 232.00 - € 462.00
Specifications
| Antigen | ZFP90 |
|---|---|
| Clone | PCRP-ZFP90-1C5 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry : 1-2 μg/million cells |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18623034
|
Novus Biologicals
NBP3-23672-20ug |
20 μg |
€ 232.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18670535
|
Novus Biologicals
NBP3-23672-100ug |
100 μg |
€ 462.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZFP90 Monoclonal antibody specifically detects ZFP90 in Human samples. It is validated for Flow CytometrySpecifications
| ZFP90 | |
| 0.2 mg/mL | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| KIAA1954, NK10, zfp-90, zinc finger protein 476, zinc finger protein 756, zinc finger protein 90 homolog, zinc finger protein 90 homolog (mouse), ZNF756 | |
| Recombinant fragment corresponding to protein domain of ZFP90 | |
| Primary | |
| Store at 4°C. |
| PCRP-ZFP90-1C5 | |
| Flow Cytometry : 1-2 μg/million cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 146198 | |
| IgG2a | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title