missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFYVE1 Monoclonal Antibody (OTI1A2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810160S
This item is not returnable.
View return policy
Description
ZFYVE1 Monoclonal antibody specifically detects ZFYVE1 in Human samples. It is validated for Western BlotSpecifications
| ZFYVE1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15437 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10483 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI1A2 | |
| Unconjugated | |
| SEC23B | |
| DFCP1; Double FYVE-containing protein 1; G630053K23; KIAA1589; mKIAA1589; phosphoinositide-binding protein SR3; PP10436; PPP1R172; protein phosphatase 1, regulatory subunit 172; SR3; TAFF1; Tandem FYVE fingers-1; tandem FYVE fingers-1 protein; Zfyve1; Zinc finger FYVE domain-containing protein 1; zinc finger FYVE-type containing 1; zinc finger protein, subfamily 2A, member 1; zinc finger, FYVE domain containing 1; ZNFN2A1 | |
| Human recombinant protein fragment corresponding to amino acids 392-641 of human SEC23B produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction