missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFYVE1 Monoclonal Antibody (OTI1A2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810160
This item is not returnable.
View return policy
Description
The FYVE domain mediates the recruitment of proteins involved in membrane trafficking and cell signaling to phosphatidylinositol 3-phosphate (PtdIns(3)P)-containing membranes. This gene encodes a protein which contains two zinc-binding FYVE domains in tandem. This protein displays a predomitly Golgi, endoplasmic reticulum and vesicular distribution. Alternatively spliced transcript variants have been found for this gene, and they encode two isoforms with different sizes.Specifications
| ZFYVE1 | |
| Monoclonal | |
| Unconjugated | |
| ZFYVE1 | |
| DFCP1, PPP1R172, SR3, TAFF1, ZNFN2A1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 53349 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI1A2 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9HBF4 | |
| ZFYVE1 | |
| Human recombit protein fragment corresponding to amino acids 1-308 of human ZFYVE1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction