Learn More
Invitrogen™ ZFYVE16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596797
Description
Immunogen sequence: MDSYFKAAVS DLDKLLDDFE QNPDEQDYLQ DVQNAYDSNH CSVSSELASS QRTSLLPKDQ ECVNSCASSE TSYGTNESSL NEKTLKGLTS IQNEKNVTGL DLLSSVDGGT SDEIQPLYMG RCSKPICDLI SDMGNLVHAT NSEEDIKKLL PDDFKSNADS LIGLDLSSVS DTPCVSSTDH DSDTVREQQN DISSELQNRE IGGIKELGIK VDTTLSDSYN YSGTENLKDK KIFNQLESIV DFNMSSALTR QSSKMFHAKD KLQHKSQPCG LLKDVGLVKE; Positive Samples: Mouse kidney; Cellular Location: Cytoplasm, Early endosome membrane, Peripheral membrane protein.
The ZFYVE16 gene encodes endofin, an endosomal protein implicated in regulating membrane trafficking. It is characterized by the presence of a phosphatidylinositol 3-phosphate-binding FYVE domain positioned in the middle of the molecule.
Specifications
| ZFYVE16 | |
| Polyclonal | |
| Unconjugated | |
| ZFYVE16 | |
| AI035632; B130024H06Rik; B130031L15; Endofin; endosomal-associated FYVE domain protein; Endosome-associated FYVE domain protein; endosome-associated FYVE-domain protein; Kiaa0305; mKIAA0305; PPP1R69; protein phosphatase 1, regulatory subunit 69; RGD1564784; ZFYVE16; Zinc finger FYVE domain-containing protein 16; zinc finger FYVE-type containing 16; zinc finger, FYVE domain containing 16 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 218441, 9765 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.54 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7Z3T8, Q80U44 | |
| ZFYVE16 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ZFYVE16 (NP_0555483). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.