missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFYVE19 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20952
This item is not returnable.
View return policy
Description
ZFYVE19 Polyclonal specifically detects ZFYVE19 in Human samples. It is validated for Western Blot.
Specifications
| ZFYVE19 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| ZFYVE19 | |
| Recombinant protein encompassing a sequence within the center region of human ZFYVE19. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 84936 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| FLJ14840, MLL partner containing FYVE domain, MPFYVE, zinc finger FYVE domain-containing protein 19, zinc finger, FYVE domain containing 19 | |
| Rabbit | |
| 52 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction