missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZFYVE26 Polyclonal Antibody

Product Code. 15700922 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15700922 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15700922 Supplier Invitrogen™ Supplier No. PA563138

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DSSKNESPPY SFVVRVPKAD EVEWILDLKE EENELVRSEF YYEQAPSASL CIAILNLHRD SIACGHQLIE HCCRLSKGLT NPEVDAGLL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.

Hereditary spastic paraplegias (HSPs) are genetically and phenotypically heterogeneous disorders. Spastic paraplegia with thinning of the corpus callosum (ARHSP-TCC) is a relatively frequent form of complicated hereditary spastic paraplegia in which mental retardation and muscle stiffness at onset are followed by slowly progressive paraparesis and cognitive deterioration. SPG15 is the second gene known to be responsible for ARHSP-TCC in the Italian population. Mutations in this gene are associated with autosomal recessive spastic paraplegia-15. SPG15 encodes a protein containing a FYVE zinc finger binding domain which is thought to target these proteins to membrane lipids through interaction with phospholipids in the membrane. SPG15 mRNA is widely distributed in human tissues, as well as in rat embryos, suggesting a possible role for this protein during embryonic development. SPG15 co-localizes partially with endoplasmic reticulum and endosome markers, suggesting a role in intracellular trafficking. Multiple isoforms of SPG15 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFYVE26
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZFYVE26
Gene Accession No. Q68DK2
Gene Alias 4930465A13; 9330197E15Rik; A630028O16Rik; FYVE domain-containing centrosomal protein; FYVE-CENT; Gm893; Kiaa0321; mKIAA0321; Spastizin; SPG15; Zfyve26; Zinc finger FYVE domain-containing protein 26; zinc finger FYVE-type containing 26; zinc finger, FYVE domain containing 26
Gene Symbols ZFYVE26
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZFYVE26. Recombinant protein control fragment (Product #RP-101111).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23503
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.