Learn More
ZG16 Mouse anti-Human, Unconjugated, Clone: 1A7B9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67389-1-IG-150UL
Description
The ZG16 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ZG16. This antibody recognizes human, mouse, pig, rat antigen. The ZG16 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| ZG16 | |
| Monoclonal | |
| Unconjugated | |
| 150 μL | |
| 653808 | |
| IgG1 |
| Western Blot, Immunohistochemistry, ELISA | |
| 1A7B9 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |