missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZGLP1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142561
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
Glucagon is initially synthesized as proglucagon, which is 180 amino acids in length and is cleaved proteolytically. It contains a region corresponding to Glucagon itself, in addition to a signal peptide, spacer regions and two Glucagon-like peptide sequences. GLP1 is encoded by the fourth exon of the six in the human Glucagon gene. It is 37 amino acids in length and has 48% amino acid sequence homology to Glucagon, and is thought to have arisen by gene duplication. The sequence of GLP1 is conserved across many species, however, indicating that it has a particular function. GLP1 is a potent insulin secretagogue, renders pancreatic beta cells 'glucose-competent' and may be useful in the treatment of diabetes mellitus.
Specifications
| ZGLP1 | |
| Polyclonal | |
| Unconjugated | |
| ZGLP1 | |
| GATA like protein 1; GATA like protein-1; GATA zinc finger domain containing 3; GATAD3; GATA-like 1; GATA-like protein 1; GATA-type zinc finger protein 1; Glp1; GLP-1; ZGLP1; zinc finger, GATA-like protein 1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 100125288 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P0C6A0 | |
| ZGLP1 | |
| Peptide with sequence C-EGVTLKFQIKPDS. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction