missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZGLP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55292-1-AP-150UL
This item is not returnable.
View return policy
Description
Glucagon is initially synthesized as proglucagon, which is 180 amino acids in length and is cleaved proteolytically. It contains a region corresponding to Glucagon itself, in addition to a signal peptide, spacer regions and two Glucagon-like peptide sequences. GLP1 is encoded by the fourth exon of the six in the human Glucagon gene. It is 37 amino acids in length and has 48% amino acid sequence homology to Glucagon, and is thought to have arisen by gene duplication. The sequence of GLP1 is conserved across many species, however, indicating that it has a particular function. GLP1 is a potent insulin secretagogue, renders pancreatic beta cells ′glucose-competent′ and may be useful in the treatment of diabetes mellitus.Specifications
| ZGLP1 | |
| Polyclonal | |
| Unconjugated | |
| ZGLP1 | |
| GATA like protein 1, GLP 1, GLP1, GLP-1, ZGLP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100009600, 100125288 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P0C6A0, Q1WG82 | |
| Zglp1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction