Learn More
ZGPAT Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
Brand:  Novus Biologicals NBP2-41146
 View more versions of this product
Product Code. 18122589
  • € 507.00 / 0.10mg
Description
ZGPAT Polyclonal specifically detects ZGPAT in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZGPAT
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
ZGPAT
Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ZGPAT. The immunogen is located within the last 50 amino acids of ZGPAT. Amino Acid Squence: CQEQRKADTHKKMTEF
0.1 mg
Primary
Multiple isoforms of ZGPAT are known to exist.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
1 mg/mL
Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL
GPATC6G patch domain-containing protein 6, GPATCH6Zinc finger and G patch domain-containing protein, KIAA1847Zinc finger CCCH domain-containing protein 9, MGC44880, ZC3H9FLJ14972, ZC3HDC9dJ583P15.3, zinc finger, CCCH-type with G patch domain, ZIPzinc finger CCCH-type with G patch domain-containing protein
Chicken
Affinity Purified
RUO
84619
Human
IgY