missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZGPAT Antibody, DyLight 755, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-41146IR
This item is not returnable.
View return policy
Description
ZGPAT Polyclonal antibody specifically detects ZGPAT in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| ZGPAT | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 755 | |
| GPATC6G patch domain-containing protein 6, GPATCH6Zinc finger and G patch domain-containing protein, KIAA1847Zinc finger CCCH domain-containing protein 9, MGC44880, ZC3H9FLJ14972, ZC3HDC9dJ583P15.3, zinc finger, CCCH-type with G patch domain, ZIPzinc finger CCCH-type with G patch domain-containing protein | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ZGPAT. The immunogen is located within the last 50 amino acids of ZGPAT. Amino Acid Squence: CQEQRKADTHKKMTEF | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 84619 | |
| Store at 4°C in the dark. | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction