missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZHX1, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00011244-D01.100uL
This item is not returnable.
View return policy
Description
Sequence: MASRRKSTTPCMVLASEQDPDLELISDLDEGPPVLTPVENTRAESISSDEEVHESVDSDNQQNKKVEGGYECKYCTFQTPDLNMFTFHVDSEHPNVVLNSSYVCVECNFLTKRYDALSEHNLKYHPGEENFKLTMVKRNNQTIFEQTINDLTFDGSFVKEENAEQAESTEVSSSGISISKTPIMKMMKNKVENKRIAVHHNSVEDVPEEKENEIKPDREEIVENPSSSASESNTSTSIVNRIHPSTASTVVTPAAVLPGLAQVITAVSAQQNSNLIPKVLIPVNSIPTYNAALDNNPLLLNTYNKFPYPTMSEITVLSAQAKYTEEQIKIWFSAQRLKHGVSWTPEEVEEARRKQFNGTVHTVPQTITVIPTHISTGSNGLPSILQTCQIVGQPGLVLTQVAGTNTLPVTAPIALTVAGVPSQNNIQKSQVPAAQPTAETKPATAAVPTSQSVKHETALVNPDSFGIRAKKTKEQLAELKVSYLKNQFPHDSEIIRLMKITGLTKGEIKKWFSDTRYNQRNSKSNQCLHLNNDSSTTIIIDSSDETTESPTVGTAQPKQSWNPFPDFTPQKFKEKTAEQLRVLQASFLNSSVLTDEELNRLRAQTKLTRREIDAWFTEKKKSKALKEEKMEIDESNAGSSKEEAGETSPADESGAPKSGSTGKICKKTPEQLHMLKSAFVRTQWPSPEEYDKLAKESGLARTDIVSWFGDTRYAWKNGNLKWYYYYQSANSSSMNGLSSLRKRGRGRPKGRGRGRPRGRPRGSKRINNWDRGPSLIKFKTGTAILKDYYLKHKFLNEQDLDELVNKSHMGYEQVREWFAERQRRSELGIELFEENEEEDEVIDDQEEDEEETDDSDTWEPPRHVKRKLSKSDDSpecifications
| ZHX1 | |
| Polyclonal | |
| No additive | |
| NM_001017926.1 | |
| ZHX1 | |
| ZHX1 (NP_001017926.1, 1 a.a. ∼ 873 a.a) full-length human protein. | |
| RUO | |
| 11244 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| zinc fingers and homeoboxes 1 | |
| - | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction