missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZHX2 Monoclonal Antibody (OTI9A5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810688S
This item is not returnable.
View return policy
Description
ZHX2 Monoclonal antibody specifically detects ZHX2 in Human samples. It is validated for Western BlotSpecifications
| ZHX2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P52435 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5439 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI9A5 | |
| Unconjugated | |
| POLR2J | |
| AFP regulator 1; AFR1; Afr-1; alpha fetoprotein regulation 1, adult; alpha-fetoprotein regulator 1; KIAA0854; LOW QUALITY PROTEIN: zinc fingers and homeoboxes protein 2; mKIAA0854; Raf; Regulator of AFP; transcription factor ZHX2; Zhx2; Zinc finger and homeodomain protein 2; zinc fingers and homeoboxes 2; zinc fingers and homeoboxes protein 2; zinc-fingers and homeoboxes 2 | |
| Full length human recombinant protein of human POLR2J produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction