missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZHX3 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-68219AF405
This item is not returnable.
View return policy
Description
ZHX3 Polyclonal antibody specifically detects ZHX3 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| ZHX3 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| FLJ93221, Sp38, zona pellucida binding protein, zona pellucida-binding protein 1, ZPBP1zona pellucida binding protein 1 | |
| The immunogen recognized by this antibody maps to a region between residue 150 and 200 of human zinc fingers and homeoboxes 3 using the numbering given in entry NP_055850.1 (GeneID 23051). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23051 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction