missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZIC Pan Polyclonal Antibody
GREENER_CHOICE

Product Code. 15837023
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15837023 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15837023 Supplier Invitrogen™ Supplier No. PA568610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 100%; Guinea Pig: 86%; Human: 100%; Mouse: 100%; Pig: 92%; Rabbit: 100%; Zebrafish: 79%.

This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. Members of this family are important during development. Aberrant expression of this gene is seen in medulloblastoma, a childhood brain tumor. This gene is closely linked to the gene encoding zinc finger protein of the cerebellum 4, a related family member on chromosome 3. This gene encodes a transcription factor that can bind and transactivate the apolipoprotein E gene. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIC Pan
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZIC3
Gene Accession No. O60481, O95409, Q15915, Q8N9L1, Q96T25
Gene Alias 1700049L20Rik; Bn; CRS6; GENA 29; heterotaxy 1; HPE5; HTX; HTX1; Ka; Ku; kumba; LOW QUALITY PROTEIN: zinc finger protein ZIC 5; Odd paired-related protein; odd-paired homolog; odd-paired related; Opa-related protein; Opr; RGD1561261; VACTERLX; Zic; Zic family member 1; Zic family member 1 (odd-paired homolog, Drosophila); Zic family member 2; Zic family member 2 (odd-paired homolog, Drosophila); Zic family member 3; Zic family member 3 (odd-paired homolog, Drosophila); Zic family member 4; Zic family member 4 (odd-paired homolog, Drosophila); Zic family member 5; zic family member 5 (odd-paired homolog, Drosophila); zic protein member 1; ZIC1; ZIC2; Zic3; ZIC4; ZIC5; zinc family member 4 protein HZIC4; zinc family member 5 protein; zinc finger protein 201; Zinc finger protein 203; zinc finger protein of the cerebellum 1; Zinc finger protein of the cerebellum 2; zinc finger protein of the cerebellum 3; Zinc finger protein of the cerebellum 4; zinc finger protein of the cerebellum 5; Zinc finger protein ZIC 1; zinc finger protein ZIC 2; Zinc finger protein ZIC 3; zinc finger protein ZIC 4; Zinc finger protein ZIC 5; zinc finger protein ZIC 5; LOW QUALITY PROTEIN: zinc finger protein ZIC 5; zinc finger protein Zic5; ZNF201; ZNF203
Gene Symbols ZIC1, ZIC2, ZIC3, ZIC4, Zic5
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human ZIC Pan.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7545, 7546, 7547, 84107, 85416
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.