Learn More
ZIC1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25004-1-AP-20UL
 View more versions of this product
Product Code. 18080626
  • € 145.00 / 20µL
Description
The ZIC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ZIC1. This antibody recognizes human, rat antigen. The ZIC1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ZIC1
500 μg/mL
Rabbit
Primary
Human, Rat
150 kDa
IgG
Western Blot, ELISA
ZIC, ZIC1, Zinc finger protein ZIC 1, ZNF201
20 μL
7545
Store at -20°C. Stable for one year after shipment.