Learn More
ZIC1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25004-1-AP-20UL
This item is not returnable.
View return policy
Description
The ZIC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ZIC1. This antibody recognizes human, rat antigen. The ZIC1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ZIC1 | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| ZIC, ZIC1, Zinc finger protein ZIC 1, ZNF201 | |
| 20 μL | |
| 7545 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |