missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZIC2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11501573
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11501573 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11501573 Supplier Invitrogen™ Supplier No. PA131023

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-31023 detects ZIC2 from mouse samples.

ZIC2 (zinc finger protein of the cerebellum 2) is a transcriptional repressor and may regulate tissue specific expression of dopamine receptor D1. In mouse, knockout of Zic1 leads to aplasia of cerebellum and skeletal abnormalities. Mutation in Zic2 and Zic3 in mouse and/or human cause holoprosencephaly and heterotaxis, respectively. Holoprosencephaly is the most common structural anomaly of the human brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIC2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene ZIC2
Gene Accession No. Q62520
Gene Alias cerebellum zinc finger protein 2; GENA 29; HPE5; Ku; kumba; LOW QUALITY PROTEIN: zinc finger protein ZIC 2; odd-paired homolog; Zic family member 2; Zic family member 2 (odd-paired homolog, Drosophila); ZIC2; Zinc finger protein of the cerebellum 2; zinc finger protein ZIC 2; zinc finger protein Zic2
Gene Symbols ZIC2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to an internal region near amino acids 180-210 of Mouse ZIC-2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22772, 361096
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.