Learn More
Invitrogen™ ZIC3 Monoclonal Antibody (2G1)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA520240
Product Code. 15508872
  • € 514.00 / 100µg
Description
Peptide Sequence: NQVHLGLRGE LFGRADPYRP VASPRTDPYA AGAQFPNYSP MNMNMGVNVA AHHGPGAFFR YMRQPIKQEL SCKWIDEAQL SRPKKSCDRT FST.
This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. This nuclear protein probably functions as a transcription factor in early stages of left-right body axis formation. Mutations in this gene cause X-linked visceral heterotaxy, which includes congenital heart disease and left-right axis defects in organs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZIC3
Monoclonal
0.5 mg/mL
PBS with no preservative; pH 7.4
O60481
Zic3
ZIC3 (NP_003404.1, 182 a.a. ~ 274 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Western Blot, Immunocytochemistry
2G1
Unconjugated
Zic3
Bn; heterotaxy 1; HTX; HTX1; Ka; RGD1561261; VACTERLX; ZIC; Zic family member 3; Zic family member 3 (odd-paired homolog, Drosophila); ZIC1; ZIC2; Zic3; Zinc finger protein 203; zinc finger protein of the cerebellum 3; Zinc finger protein ZIC 3; ZNF201; ZNF203
Mouse
Affinity chromatography
RUO
7547
-20°C, Avoid Freeze/Thaw Cycles