Learn More
ZIC3 Mouse anti-Human, Clone: OTI6F7, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF808167
Beschreibung
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. This nuclear protein probably functions as a transcription factor in early stages of left-right body axis formation. Mutations in this gene cause X-linked visceral heterotaxy, which includes congenital heart disease and left-right axis defects in organs.Spezifikation
| ZIC3 | |
| Monoclonal | |
| Unconjugated | |
| ZIC3 | |
| HTX, HTX1, VACTERLX, ZNF203 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7547 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI6F7 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O60481 | |
| ZIC3 | |
| Human recombit protein fragment corresponding to amino acids 242-467 of human ZIC3 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Das Fisher Scientific Encompass-Programm bietet Artikel an, die nicht Teil unseres Vertriebsportfolios sind. Diese Produkte verfügen in der Regel nicht über Bilder oder detaillierte Beschreibungen. Wir sind jedoch bestrebt, Ihr Einkaufserlebnis zu verbessern. Bitte verwenden Sie das untenstehende Formular, um uns Feedback zum Inhalt dieses Produkts zu geben.