missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zika Virus IgG Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-61131
This item is not returnable.
View return policy
Description
Zika Virus IgG Polyclonal specifically detects Zika Virus IgG in Viral samples. It is validated for ELISA.
Specifications
| Zika Virus IgG | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Zika Capsid antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human Zika Capsid. Amino Acid Squence: RIINARKEKKRRGADTS | |
| 0.1 mg | |
| Zika | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| ELISA | |
| 1 mg/mL | |
| ELISA 1:100-1:2000 | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction