missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zika Virus IgG Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 178.00 - € 507.00
Specifications
| Antigen | Zika Virus IgG |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100-1:2000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18221612
|
Novus Biologicals
NBP2-61133 |
0.1 mg |
€ 507.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18636697
|
Novus Biologicals
NBP2-61133-0.025mg |
0.025 mg |
€ 178.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zika Virus IgG Polyclonal specifically detects Zika Virus IgG in Viral samples. It is validated for ELISA.Specifications
| Zika Virus IgG | |
| ELISA 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| PBS with 0.02% Sodium Azide | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Virus | |
| Zika Envelope antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human Zika Envelope. Amino Acid Squence: QGEAYLDKQSDTQYV | |
| Primary | |
| Zika |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title