missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zika Virus IgG Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-61132R
This item is not returnable.
View return policy
Description
Zika Virus IgG Polyclonal antibody specifically detects Zika Virus IgG in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Zika Virus IgG | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Zika Membrane antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human Zika Membrane. Amino Acid Squence: LPSHSTRKLQTRSQTWLE | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| DyLight 550 | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction