missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zika Virus NS5 (strain H/PF/2013) Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5143441

Product Code. 17870596

  • € 464.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Store as concentrated solution. Centrifuge briefly prior to opening vial. Positive control: Zika-infected Vero.

Zika Virus nonstructural protein 5 (NS5) interacts with the host retinoic acid-inducible gene I (RIG-I), which is an essential signaling molecule for defending pathogen infections. NS5 subsequently represses K63-linked polyubiquitination of RIG-I, attenuates the phosphorylation and nuclear translocation of interferon regulatory factor 3 (IRF3), and inhibits the expression and production of interferon-beta (IFN-beta), thus restricting the RIG-I signaling pathway.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Zika Virus NS5 (strain H/PF/2013)
Polyclonal
Unconjugated
NS5; NS5 protein
Recombinant protein encompassing a sequence within the N-terminus region of Zika virus NS5 protein (Zika virus (strain H/PF/2013)). The exact sequence is proprietary.
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunoprecipitation, Western Blot, Immunocytochemistry
0.78 mg/mL
PBS with 20% glycerol and 0.025% Proclin 300; pH 7
Rabbit
Antigen affinity chromatography
RUO
Virus
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.