missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zika Virus (SPH2015) NS1 Antibody (15), Janelia Fluor™ 646 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05871JF646
This item is not returnable.
View return policy
Description
Zika Virus (SPH2015) NS1 Monoclonal antibody specifically detects Zika Virus (SPH2015) NS1 in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Zika Virus (SPH2015) NS1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| Zika NS1, ZIKV NS1, ZIKV-NS1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Zika virus (ZIKV) (strain Zika SPH2015) ZIKV-NS1 (ALU33341.1; Val796-Leu1157). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| 15 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction