missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zika Virus (SPH2015) NS1 Antibody (201), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-05872-100ul
This item is not returnable.
View return policy
Description
Zika Virus (SPH2015) NS1 Monoclonal antibody specifically detects Zika Virus (SPH2015) NS1 in Virus samples. It is validated for ELISA
Specifications
| Zika Virus (SPH2015) NS1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA | |
| 201 | |
| ELISA 1:5000-1:10000 | |
| Zika NS1, ZIKV NS1, ZIKV-NS1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Zika Virus (Accession#: ALU33341.1; Val796-Leu1157). | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction