Learn More
Invitrogen™ Zilovertamab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558395
 View more versions of this product
Product Code. 30283659
  • € 477.00 / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Zilovertamab targets and binds to the receptor tyrosine kinase-like orphan receptor 1 (ROR1), inhibiting its signaling and promoting cancer cell death.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Zilovertamab Humanized
Recombinant Monoclonal
Unconjugated
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Protein A