missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Synapsin 1 Recombinant Rabbit Monoclonal Antibody (3I9R10)

Description
Immunogen sequence: EDKQLIVELV VNKMAQALPR QRQRDASPGR GSHGQTPSPG ALPLGRQTSQ QPAGPPAQQR PPPQGGPPQP GPGPQRQGPP LQQRPPPQGQ QHLSGLGPPA.
SYN1 (synapsin 1) is a member of the synapsin gene family. Synapsins encode neuronal phosphoproteins that associate with the cytoplasmic surface of synaptic vesicles. Family members are characterized by common protein domains and are implicated in synaptogenesis and the modulation of neurotransmitter release, suggesting a potential role in several neuropsychiatric diseases. Synapsin 1 plays a role in regulation of axonogenesis and synaptogenesis. It serves as a substrate for several different protein kinases and phosphorylation may function in the regulation of this protein in the nerve terminal. Mutations in this gene may be associated with X-linked disorders with primary neuronal degeneration such as Rett syndrome. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
Specifications
| Antigen | Synapsin 1 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Recombinant Monoclonal |
| Clone | 3I9R10 |
| Concentration | 0.57 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | SYN1 |
| Gene Accession No. | O88935, P09951 |
| Gene Alias | Brain protein 4.1; SYN1; Syn-1; SYN1a; SYN1b; Syn1-S; synapsin 1; synapsin I; synapsin Ia; synapsin Ib; synapsin-1; SYNI; truncated synapsin 1-S |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?