Learn More
Invitrogen™ Ziltivekimab Recombinant Monoclonal Antibody
GREENER_CHOICE
Product Code. 30283165
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30283165 100 μg 100µg
30283158 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30283165 Supplier Invitrogen™ Supplier No. MA559601
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Ziltivekimab is a proprietary anti-interleukin-6 ligand monoclonal antibody (anti-IL6 mAb), targeting residual inflammatory cardiovascular risk in patients living with advanced chronic kidney disease (CKD).
TRUSTED_SUSTAINABILITY
Specifications
Antigen Ziltivekimab
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Gene Alias COR-001
Host Species Human
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1