missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZIM2 Antibody (OTI6D12), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74928FR
This item is not returnable.
View return policy
Description
ZIM2 Monoclonal specifically detects ZIM2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ZIM2 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| ZIM2 | |
| Recombinant protein fragment corresponding to amino acids 1-150 and 428-527 of human ZIM2 (NP_056178) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI6D12 | |
| Western Blot, Immunohistochemistry | |
| zinc finger imprinted 2, zinc finger, imprinted 2, ZNF656Zinc finger protein 656 | |
| Mouse | |
| Affinity Purified | |
| modENCODE Antibodies | |
| 23619.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction