missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZIM2 Monoclonal Antibody (OTI5C7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501904
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
In human, ZIM2 and PEG3 (GeneID:5178) are two distinct genes that share a set of 5′ exons and have a common promoter, and both genes are paternally expressed. Alternative splicing events connect the shared exons either with the remaining 4 exons unique to ZIM2, or with the remaining 2 exons unique to PEG3. This is in contrast to mouse and cow, where ZIM2 and PEG3 genes do not share exons in common, and the imprinting status of ZIM2 is also not conserved amongst mammals. Alternatively spliced transcripts encoding the same protein have been found for the human ZIM2 gene.Tekniske data
| ZIM2 | |
| Monoclonal | |
| 0.6 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NZV7 | |
| ZIM2 | |
| Human recombit protein fragment corresponding to amino acids 1-150 and 428-527 of human ZIM2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI5C7 | |
| Unconjugated | |
| ZIM2 | |
| ZNF656 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23619 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?